▸
Bottles, Jars and Jugs
▸
Jars
▸
Storage Jars
▸
Screw Neck Cylindrical Clear Glass Jar
Actual product may vary
Fisherbrand
Screw Neck Cylindrical Clear Glass Jar
Can be used for soil samples.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15692757
50 mL
Catalog number
FKS15692757
Price (SGD)
378.00
Pack
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
378.00
Pack
Add to cart
50mL
Screw Neck ND40
Note:
Lid is sold separately.
Specifications
Capacity (Metric)
50 mL
Color
Clear
Material
Glass
No. per Pack
88
Packaging
PP-Box
Clarity
Clear
Shape
Cylindrical
Height (Metric)
69 mm
Product Type
Jar
Diameter (Metric)
44 mm
Closure Type
40mm Screw Cap (Sold separately)