Lab Thermometer
Lab Thermometer
Actual product may vary
Fisherbrand
Lab Thermometer
General purpose laboratory thermometer without calibration certificate.
Have Questions?
Catalog NumberTemperature Range (Metric)SubdivisionImmersion TypeLength (Metric)
FKS15683204-100°C to +50°C1°CTotal Immersion300 mm
Catalog number FKS15683204
Price (SGD)
362.00
EA
Add to cart
Temperature Range (Metric):
-100°C to +50°C
Subdivision:
1°C
Immersion Type:
Total Immersion
Length (Metric):
300 mm
Price (SGD)
362.00
EA
Add to cart
Filled with a brightly colored, mercury-free spirit. Brannan™ Partial Immersion Spirit Filled Thermometers use specially selected filling materials covering temperatures down to -200°C and feature a non-roll cap with suspension ring.
  • Lab thermometer
  • Safe non-mercury filling
  • Non-roll end cap
  • Supplied as standard in protective individual, resealable telescopic tube
  • Made in UK
Specifications
Diameter (Metric) Probe7 mm
For Use With (Application)General use
Immersion TypeTotal Immersion
Length (Metric)300 mm
MaterialGlass
Thermometer FillSpirit
Product TypeLow Temperature Thermometer
Max. Temperature (Metric)50°C
Min. Temperature (Metric)-100°C
Temperature Range (Metric)-100°C to +50°C
Subdivision1°C
Width (Metric)1.2 cm
Unit SizeEA