SteriBags™ Heat Seal Closure Pouches
Product Image
Fisherbrand
SteriBags™ Heat Seal Closure Pouches
Manufactured from high quality wet strength MG bleached kraft paper. Synergy Healthcare™ SteriBags™ Heat Seal Closure Pouches are double-folded for added integrity.
Have Questions?
Catalog NumberDimensions (L x W)
FKS15671944380 x 180 mm
Catalog number FKS15671944
Price (SGD)
425.00
PK
Add to cart
Dimensions (L x W):
380 x 180 mm
Price (SGD)
425.00
PK
Add to cart
  • Conforms to BS 6257
  • Double folded bottom for extra integrity
  • Opening along width
  • For use in autoclaves and gamma sterilizers
Specifications
DescriptionSterilization pouch with heat seal closure, code H
AutoclavableAutoclavable
Certifications/ComplianceConforms to BS 6257; Code H
ColorWhite
For Use With (Application)For use in porous load pre-vacuum autoclaves as well as gamma sterilizers
MaterialKraft Paper
Quantity500/Pk.
Dimensions (L x W)380 x 180 mm
Dimensions (L x W x H)380 x 180 x 95 mm (L x W x D)
Length (Metric)380 mm
Width (Metric)180 mm
Product TypeSterilization Pouch
Unit SizePK