Microflex™ 93-260 Chemical-Resistant Disposable Glove
Microflex™ 93-260 Chemical-Resistant Disposable Glove
Actual product may vary
Microflex™ 93-260 Chemical-Resistant Disposable Glove
Microflex™ 93-260 Chemical-Resistant Disposable Glove
Microflex™ 93-260 Chemical-Resistant Disposable Glove
Fisherbrand
Microflex™ 93-260 Chemical-Resistant Disposable Glove
Provides thin yet tough chemical protection with unparalleled comfort. Microflex™ 93-260 Disposable Gloves have a solvent resistant exterior, an acid and base resistant middle layer and an easy donning interior layer.
Have Questions?
Catalog NumberSize
FKS15653074Large
Catalog number FKS15653074
Price (SGD)
-
Size:
Large
  • Three layer design for superior protection against harsh chemicals including acids, solvents and bases
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
  • Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone free formulation and processing ensure better product protection
SIZE CHART
  • XS – 5.5 to 6
  • S – 6.5 to 7
  • M – 7.5 to 8
  • L – 6.5 to 9
  • XL – 9.5 to 10
  • 2XL – 10.5 to 11
Specifications
Certifications/ComplianceASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III
ColorGreen, Blue
DisposableSingle-use
HandAmbidextrous
MaterialNitrile, Neoprene
PowderedPowder Free
Product TypeDisposable Gloves
SizeLarge
Thickness (Metric)0.198 mm
Finger StyleTextured
Glove Cuff StyleExtended, Beaded
Model93-260
Packaging TypeDispenser box
Puncture Rating0.65 AQL
Quantity50Pack
Resistant ToHarsh chemicals including acids, solvents and bases
Tensile Strength22 MPa
Length (Metric)300 mm
Width (Metric)111 mm
Unit SizePK