Azo™ Gusseted Reels
Azo™ Gusseted Reels
Actual product may vary
Fisher Catalogue
Azo™ Gusseted Reels
High quality preformed, heat sealable, peelable sterile barrier packaging. Synergy Healthcare™ Azo™ Gusseted Reels are suited to the sterilization of surgical instruments, laboratory equipment or small to medium sized sets using steam or Ethylene Oxide.
Have Questions?
Catalog NumberDimensions (L x W)Quantity
FKS15631974100 m x 100 mm1 Ea.
Catalog number FKS15631974
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
100 m x 100 mm
Quantity:
1 Ea.
  • Protects its contents from deterioration, damage and contamination
  • Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
AutoclavableAutoclavable
Certifications/ComplianceEN868-5 and ISO11607-1
ColorBlue
MaterialPolyester, Polypropylene
SterilityNon-sterile
Quantity1 Ea.
Sterilization MethodSteam and EO
Dimensions (L x W)100 m x 100 mm
Length (Metric)200 m
Product TypeSterilization Reel
Unit SizePK