SPRIselect™
Product Image
SPRIselect™
Purify DNA fragments of the desired size for library preparation. Beckman Coulter™ SPRIselect™ chemistry speeds and simplifies nucleic acid size selection using paramagnetic bead-based chemistry for next-generation sequencing.
Have Questions?
Catalog NumberQuantity
FKS1560583860mL
Catalog number FKS15605838
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
60mL
Proprietary SPRI paramagnetic bead-based chemistry provides:
  • Tunable fragment size selection to suit specific applications
  • Predictable, consistent size selection between runs and reagent lots
  • Compatibility with manual and automated processing
  • Customizable size selection: from 150 to 800 base pairs
  • Suitable for manual or automated workflows; can be run on Biomek platforms
Specifications
FormLiquid
FormatLiquid
For Use With (Equipment)Automated processing systems, e.g. Biomek
pH8.0 - 8.4
Quantity60mL
For Use With (Application)Standard PCR
Sample TypeNucleic Acid
Size (Final Product)Tunable size selection from 150 to 800 base pairs
Unit SizeEA
Contents & Storage
15-30°C