Beatus Bottle Top Dispenser
Beatus Bottle Top Dispenser
Actual product may vary
Fisherbrand
Beatus Bottle Top Dispenser
Robust and reliable dispensing; Fisherbrand™ Beatus Bottle Top Dispenser has re-circulation valve for re-directing the liquid into the bottle for bubble free dispensing without any loss of reagent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)IncrementsAccuracyModel
FKS155988911 to 10 mL0.2 mL0.6% (+/-0.060)BEAT-10
FKS155389010.5 to 5 mL0.1 mL0.6% (+/-0.030)BEAT-5
Catalog number FKS15598891
Price (SGD)
656.00
EA
Add to cart
Volume (Metric):
1 to 10 mL
Increments:
0.2 mL
Accuracy:
0.6% (+/-0.060)
Model:
BEAT-10
Price (SGD)
656.00
EA
Add to cart
  • With re-circulation valve
  • Adjustable delivery nozzle to facilitate dispensing ease in all laboratory conditions
  • Specially designed volume adjustment knob, 180° rotation for easy and effortless volume setting
  • Specially designed adaptor allows 360° rotation providing full flexibility
  • Springless, PTFE valve manifold ensures high chemical compatibility and jamming free valve function
  • PTFE piston with ETP O-ring ensures high chemical compatibility and wiper smooth, effortless piston movement
  • Adaptors to fit most of the laboratory reagent bottles
  • Calibration tool provided for convenient and quick in-lab user re-calibration
Specifications:
  • Piston: PTFE and ETP
  • Valve housing: PFA
  • Re-Circulation valve housing: PFA
  • Valve assembly: Borosilicate glass and seat
  • Discharge assembly: PTFE
  • Delivery and Inlet tube: FEP
  • Compatibility: With all reagents except HF
Specifications
Accuracy0.6% (+/-0.060)
Product TypeBottletop Dispenser
AutoclavableAutoclavable (121°C, 15psi for 10-15 min.)
Certifications/ComplianceISO 8655 standards
MaterialBorosilicate Glass
Volume (Metric)1 to 10 mL
Increments0.2 mL
ModelBEAT-10
Unit SizeEA