150 Watt Hand Held Homogenizer
Actual product may vary
Fisherbrand
150 Watt Hand Held Homogenizer
Process samples reliably, quickly and efficiently. Fisherbrand™ 150 Watt Hand Held Homogenizer is a high performance, light weight tissue homogenizer.
Have Questions?
Catalog NumberPlug Type
FKS15575809UK Plug
Catalog number FKS15575809
Price (SGD)
3,500.00
Each
Plug Type:
UK Plug
Price (SGD)
3,500.00
Each
  • Designed for comfortable hand-held use/can be mounted to motor stand for lengthier processing
  • Interchangeable quick connect rotor stator generator probes provides flexibility to process samples from 200μL to 100mL
  • Stainless steel probes (5mm - 10mm diameter) are designed for long life and are easy to disassemble for cleaning
  • Plastic probes virtually eliminate risk of cross contamination and carryover between samples, can be cleaned for reuse in most of the cases
  • Speed: Variable from 5,000 to 35,000rpm
  • Power: 150w max
  • Sound: <72dB
Specifications
Product TypeHomogenizer
Speed5000 to 35,000 rpm
Electrical Requirements220V UK Cord, 150W
Diameter (Metric)6.35 cm
Height (Metric)22.8 cm
For Use With (Application)Tissue homogenization, PCR analysis, cell disruption, RNA/DNA and protein extractions, and emulsions and suspensions
Packaging Quantity1 Ea.
Plug TypeUK Plug
Weight (Metric)466 g
Unit SizeEach