Check Order Status
Grade 601 Cellulose General Purpose Filter Paper
Actual product may vary
Fisherbrand
Grade 601 Cellulose General Purpose Filter Paper
Used in analytical works in laboratories. Fisherbrand™ Grade 601 Cellulose General Purpose Filter Paper has medium filtration velocity.
Have Questions?
Change view
Catalog Number
Format
Diameter (Metric)
Dimensions (L x W)
FKS15535689
Disc
150 mm
FKS15525689
Disc
125 mm
FKS11705365
Sheet
570 x 460mm
3 Options
Catalog number
FKS15535689
Price (SGD)
-
Format:
Disc
Diameter (Metric):
150 mm
Applications:
Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifications
Format
Disc
Material
Cellulose
Particle Retention
5 to 13 μm
Pore Size
5 to 13 μm
For Use With (Application)
Filter Paper used in analytical works in laboratories, medium filtration velocity
Grade
601
Quantity
100/Pk.
Thickness (Metric)
0.16 mm
Diameter (Metric)
150 mm