Mini Tube Rotator
Mini Tube Rotator
Actual product may vary
Mini Tube Rotator
Mini Tube Rotator
Mini Tube Rotator
Can be utilized across a broad spectrum of scientific disciplines,including, but not limited to, molecular biology, clinical, and biochemistry
Have Questions?
Catalog NumberPlug TypeCertifications/Compliance
FKS15534080EU, UK, CN PlugCE
Catalog number FKS15534080
Price (SGD)
1,434.00
EA
Add to cart
Plug Type:
EU, UK, CN Plug
Certifications/Compliance:
CE
Price (SGD)
1,434.00
EA
Add to cart
  • Small footprint
  • Digital RPM Display
  • Maintenance free motor with quiet operation
  • Paddles are easily adjustable/interchanged - Each paddle can be adjusted from 0-90 degrees
  • Achieve more effective mixing with “agitation” mode
  • Agitation will occur at every 40 degrees in the rotation cycle
  • Combination of rotation and oscillation provides more effective mixing
  • Extra paddles available
Specifications
Speed Control Accuracy±1 rpm
Speed Range10 to 40 rpm
Voltage100/240 V
Temperature Range (Metric)4°C to 40°C
HoldsTubes
Certifications/ComplianceCE
Plug TypeEU, UK, CN Plug
No. of Positions
  • 84 x 0.5/0.8mL tubes
  • 60 x 1.5/2.0mL tubes
  • 28 x 5/7mL tubes
  • 28 x 10/15mL tubes
  • 6 x 50mL tubes
Warranty and Service Offering2 Years
Unit SizeEA