Multi-Purpose Tube Rotators
Actual product may vary
Fisherbrand
Multi-Purpose Tube Rotators
Versatile, robust, multi-purpose rotator suitable for most mixing needs
Have Questions?
Catalog NumberPlug TypeCertifications/Compliance
FKS15524080EU, UK, CN PlugCE
Catalog number FKS15524080
Price (SGD)
2,053.00
Each
Plug Type:
EU, UK, CN Plug
Certifications/Compliance:
CE
Price (SGD)
2,053.00
Each
  • Digital microprocessor control 
  • Five carousel options 
  • Adjustable speed provides gentle or vigorous mixing action 
  • Adjustable angled head offers mixing angles from 95 to 180° 
  • Timer features both time up and time down counting modes 
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Wide range voltage: AC100-240V. 
  • Audible and visual alarm: In timed mode, when the time reaches zero, alarm will sound 5 times, then the time window shows“End“ and flickers all the time. Alarm can be stopped by touch any key. 
Specifications
Rotating AngleCarousel Angle: 95°- 180°(Six steps : 95°, 112°, 129°, 146°, 163°, 180º)
Speed Range5 to 80 rpm
Voltage100/240 V
HoldsTubes, Culture Tubes with Drum Carousel
Tube Capacity50 mL
Product TypeTube Carousel
Certifications/ComplianceCE
Plug TypeEU, UK, CN Plug
Dimensions (L x W x H)415 x 293 x 464 mm
No. of Positions
  • 50 x 9-11mm tubes
  • 30 x14-18mm tubes
  • 15 x 27-34mm tubes
  • 10 x 10-15ml and 20 x  5-7ml tube
  • 64 x 18.5mm culture tubes (with drum carousel)
  • 64 x 14mm culture tubes (with drum carousel)
Timer Range0 min. to 99:59 hr.
Warranty and Service Offering2 Years
Unit SizeEach