accumet™ AE6 Portable pH/ORP Meter
accumet™ AE6 Portable pH/ORP Meter
Actual product may vary
Fisherbrand
accumet™ AE6 Portable pH/ORP Meter
The AE6 Portable pH Meters are ideal for quick, accurate conductivity or TDS checks in laboratories, field and schools.
Have Questions?
Catalog NumberType
FKS15523653Meter with 3-in-1 Probe
Catalog number FKS15523653
Price (SGD)
1,814.00
EA
Add to cart
Type:
Meter with 3-in-1 Probe
Price (SGD)
1,814.00
EA
Add to cart
Fisherbrand accumet AE6 Portable pH Meter, Includes: pH/Temp electrode, Accuracy: +/- 0.01pH, +/-0.5 deg.C, +/- 1mV, Range: -1000 to 1000 mV, 0 to 14, Resolution: 0.01, 1mV, 0.1 deg.C, Battery Type: 4 x AAA
  • Multipoint calibration (up to 5 points) with auto-buffer recognition
  • Choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • For analyzing water in pools and spas, lithographic processes, boiler and cooling tower water analysis, environmental and agricultural purposes, and all types of quality assurance and water quality testing
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold function momentarily freezes reading for easy viewing and recording
  • Auto-power off saves battery power after 17 minutes of non-use
  • Self-diagnostic with message codes for easy troubleshooting
  • Large custom LCD provides optimum viewing, even at a distance
  • Rubber boot shields protect from accidental knocks and feature sturdy, built-in stand for easy benchtop operation
  • Optional rugged carrying case provides handy on-site meter calibration and measurement
Specifications
Accuracy (Temperature)±0.5°C
Accuracy (mV)±1
Accuracy (pH)±0.01
IncludesAE6 pH/ORP Meter, Rubber Armor/Stand, 3-in-1 pH/Temperature Combination Electrode ECFC7352901B and pH Carrying Kit
For Use With (Application)Water Quality Testing
Calibration1.68, 4.01, 7, 10.01, 12.45
Range (pH)0 to 14
Resolution (pH)0.01
Range (mV)-1000 to +1000 mV
Resolution (mV)1 mV
Calibration Points1 to 5
Temperature Range (Metric)0°C to 100°C
Resolution (Temperature)0.1°C
ModelAE6
Temperature CompensationAutomatic/Manual (0°C to 100°C)
Battery TypeAAA
TypeMeter with 3-in-1 Probe
Unit SizeEA