Semperguard™ Nitrile Xpert Powder Free Gloves
Semperguard™ Nitrile Xpert Powder Free Gloves
Actual product may vary
Semperguard™ Nitrile Xpert Powder Free Gloves
Semperguard™ Nitrile Xpert Powder Free Gloves
Fisherbrand
Semperguard™ Nitrile Xpert Powder Free Gloves
Blue nitrile gloves are powder free and resistant to skin irritants.
Have Questions?
Catalog NumberSize
FKS15513536Large
Catalog number FKS15513536
Price (SGD)
-
Size:
Large
  • PPE Category III complex risks
  • Excellent resistance to skin irritants
  • Suitable for food contact
  • Powder free
  • Chemical resistant
  • No silicone added
Specifications
Certifications/ComplianceCE Class I; EN 420, EN 374, EN 455; PPE Category III
ClassificationCategory III
DescriptionSemperguard™ Nitrile Xpert Powder Free Gloves
ColorBlue
DisposableSingle-use
Exterior FinishPartially Textured
HandAmbidextrous
Latex FreeYes
MaterialNitrile
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeLarge
SterilityNon-sterile
Thickness (Metric)0.26 mm
For Use With (Application)Pharmaceutical and laboratory applications
Glove Cuff StyleRolled
Quantity100/Pk.
Length (Metric)24 cm
Unit SizePK