Rubber Cover
Rubber Cover
Actual product may vary
Fisher Catalogue
Rubber Cover
Fits on 76.2mm Replacement Platform. Scientific Industries SI™ Rubber Cover is used with Vortex-Geinie 1 and Vortex-Genie 2 family of mixers and Vortex-Genie Pulse.
Have Questions?
Catalog number FKS15507335
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • For use with Vortex-Genie family of products
  • Platform attachment required
Specifications
MaterialRubber
For Use With (Equipment)Vortex-Genie 1 and Vortex-Genie 2 family of mixers, Vortex-Genie Pulse
RequiresReplacement Platform 15547315
Unit SizeEA