850 Homogenizer
850 Homogenizer
Actual product may vary
Fisherbrand
850 Homogenizer
The Fisherbrand™ 850 Homogenizer is specifically designed to disrupt, emulsify or homogenize a wide range of samples in volumes ranging from 0.2mL to 5L. 
Have Questions?
Catalog number FKS15505819
Price (SGD)
9,584.00
EA
Add to cart
Price (SGD)
9,584.00
EA
Add to cart
  • Powerful 850w motor
  • Integrated digital speed control, displays speed and run time
  • Variable speed control from 5,000 to 28,000rpm
  • Compatible with stainless steel and disposable generator probes
  • Processing range from 0.2mL to 5L
  • Includes motor clamp
  • Stand (baseplate and post) is sold separately; see Cat. No. 15535819
Specifications
Product TypeHomogenizer
Speed5000 to 28,000 rpm
Electrical Requirements220 V, 850 W, EU/UK Cord
Volume Range (Metric)0.2 mL to 5 L
Diameter (Metric)6.9 cm
Height (Metric)21.5 cm
For Use With (Application)Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
IncludesHomogenizer, Motor Clamp. (Stand is sold separately, 15535819.)
Packaging Quantity1 Ea.
Plug TypeEU/UK Plug
Weight (Metric)1.6 kg
Unit SizeEA