Porcelain Plate For Desiccators
Porcelain Plate For Desiccators
Actual product may vary
Fisherbrand
Porcelain Plate For Desiccators
For use with desiccators
Have Questions?
Catalog NumberFor Use With (Equipment)Diameter (Metric) Outer
FKS15504655Desiccator (Dia.: 300mm)280 mm
Catalog number FKS15504655
Price (SGD)
115.00
EA
Add to cart
For Use With (Equipment):
Desiccator (Dia.: 300mm)
Diameter (Metric) Outer:
280 mm
Price (SGD)
115.00
EA
Add to cart
  • For use with desiccators
Specifications
Diameter (Metric) Outer280 mm
Product TypePlate
For Use With (Equipment)Desiccator (Dia.: 300mm)
Unit SizeEA