AccuDuo Cartridge
AccuDuo Cartridge
Actual product may vary
Fisherbrand
AccuDuo Cartridge
Ensures optimum quality of the final product water. Fisher Scientific™ AccuDuo Cartridges provide dual column filtration for Accu20, Accu100, and Accu500 water systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
FKS15499729U Pack w/ 0.2μm PES Capsule FilterAccu20, Accu500
FKS15449719P PackAccu20
Catalog number FKS15499729
Price (SGD)
2,130.00
EA
Add to cart
Type:
U Pack w/ 0.2μm PES Capsule Filter
For Use With (Equipment):
Accu20, Accu500
Price (SGD)
2,130.00
EA
Add to cart
  • Includes two sets of dual-column purification cartridges
  • Cartridges work sequentially to ensure thorough removal of ions and organics in water
Specifications
For Use With (Equipment)Accu20, Accu500
TypeU Pack w/ 0.2μm PES Capsule Filter
For Use With (Application)Ultrapure water for critical laboratory applications
Unit SizeEA