Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Long, tapered outside contour minimizes chipping
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1549613350 mL
FISFB5002525 mL
FISFB500125125 mL
FISFB5005050 mL
FISFB50010001000 mL
FISFB500500500 mL
FISFB500250250 mL
FISFB50020002000 mL
FISFB50040004 L
Catalog number FKS15496133
Price (SGD)
210.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
210.00
EA
Add to cart
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Closure MaterialRubber
DescriptionTop style: Heavy duty tooled with capacity scale
Diameter (English) Outer1.96 in.
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Graduation Range20 to 50 mL
Graduations10 mL
Height (English)3.07 in.
Height (Metric)7.8 cm
No. of Positions1
Writing SurfaceMatte Finish Ceramic Enamel Marking
DisposableReusable
Mouth StyleNarrow
Diameter (Metric) Outer50 mm
Closure Size1
Unit SizeEA