Accu100 Ultrapure Water System
Accu100 Ultrapure Water System
Actual product may vary
Fisherbrand
Accu100 Ultrapure Water System
Ultrapure water for critical laboratory applications. Fisherbrand™ Accu100 Ultrapure Water System used to produce Type I ultrapure water from RO, distilled or deionized water.
Have Questions?
Catalog NumberType
FKS15489749Ultrapure Water Purification System
Catalog number FKS15489749
Price (SGD)
16,552.00
EA
Add to cart
Type:
Ultrapure Water Purification System
Price (SGD)
16,552.00
EA
Add to cart
Independent Ultrapure water dispenser
  • Remote dispenser has built-in high precision resistivity sensor with cell constant of 0.01cm-1 to measure quality of ultrapure water just prior to dispensing point
  • Volumetric dispensing and calibration
  • Handle is 360° rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Multi-functional control panel
  • Volumetric dispensing and calibration
  • Alarms and maintenance info on large color LCD screen
  • Real-time monitoring of system status: Product water quality/resistivity and Cartridge/UV lamp remaining service life
AccuQuatro U Pack
  • Patented large capacity ultrapurification cartridge
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
DescriptionTOC: 0 - 15psi (0 - 1 bar)
Bacterial Content<0.1 CFU/mL
Feedwater SourceDistilled, DI, RO, EDI
Flow Rate1.0 to 1.5 L/min. at 25°C
Resistivity18.2 Mohm.cm at 25°C
TypeUltrapure Water Purification System
ModelAccu100
Particle Size<1/mL
Temperature Range (Metric)5°C to 45°C
Length (English)8.26 in.
Length (Metric)21 cm
Height (English)17.3 in.
Height (Metric)44 cm
Depth (English)13.7 in.
Depth (Metric)35 cm
Weight (English)13.2 lb.
Weight (Metric)6 kg
Unit SizeEA