Accu20 Ultrapure Water System
Accu20 Ultrapure Water System
Actual product may vary
Fisherbrand
Accu20 Ultrapure Water System
Fisherbrand™ Accu20 Ultrapure Water System used to produce Type I ultrapure water and RO water on demand from tap water directly.
Have Questions?
Catalog NumberType
FKS15489689ACCU20 Ultrapure Water System
Catalog number FKS15489689
Price (SGD)
21,120.00
EA
Add to cart
Type:
ACCU20 Ultrapure Water System
Price (SGD)
21,120.00
EA
Add to cart
  • Compact system provides easy way to produce ultrapure water or RO water on demand from tap water directly with no additional pre-treatment filters
  • Equipped with safeguard AccuDuo P pack cartridge and dual-column AccuDuo U Pack ultrapurification cartridge to ensure optimum quality of final product water
  • No external storage tank required, which eliminate risk of water degration by contaminants from container or tubing
  • Fully automatic controlled and maintained system contains automatic cleaning cycles and selfmaintenance functions for RO membrane, which increases lifetime and efficiency of RO membranes as well as cartridges
  • Automatic hourly system recirculation in loop prevents bacterial growth and maintain water purity at all times
  • System status and key quality parameters are clearly displayed on large LCD screen for easy system performance monitoring
Applications:
  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.
Specifications
ConductivityRO water at 25°C: Typically <20μS/cm
Bacterial Content<0.1 CFU/mL (with a 0.2 μm final filter or terminal ultrafiltration cartridge )
Flow RateRO water (at 25°C ): >0.5L/min.; Ultrapure water ( Type I ) (at 25°C ): >0.5L/min.
Resistivity18.2 Mohm.cm
TOC Monitor with UV Intensity Monitoring<10 ppb (<5 ppb with a dual wavelength UV lamp)
TypeACCU20 Ultrapure Water System
Feedwater Pressure15 to 90 psi (1 to 6 bar )
ModelAccu20
Particle Size<1/mL (with a 0.2μm final filter or terminal ultrafiltration cartridge)
Temperature Range (Metric)5°C to 45°C
Length (English)76.2 in.
Length (Metric)30 cm
Height (English)18.8 in.
Height (Metric)48 cm
Depth (English)20 in.
Depth (Metric)51 cm
Unit SizeEA