Eutech™ KCl Calibration Solution
Product Image
Fisherbrand
Eutech™ KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConductivity
FKS15488972480 mL12.88 mS/cm
ECCON84BT480 mL84 μS/cm
ECCON1118BT480 mL111.8 mS/cm
Catalog number FKS15488972
Price (SGD)
51.00
EA
Add to cart
Quantity:
480 mL
Conductivity:
12.88 mS/cm
Price (SGD)
51.00
EA
Add to cart
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 60mL or 480 ml durable plastic bottles
  • Economical and convenient to use
Specifications
Description12.88 mS/cm KCl calibration solution
Conductivity12.88 mS/cm
For Use With (Equipment)Conductivity Meters and Electrodes
Solution TypeCalibration Solution
TypeConductivity Standard
Quantity480 mL
Unit SizeEA