Straight Polypropylene Container with Screw Cap
Straight Polypropylene Container with Screw Cap
Actual product may vary
Straight Polypropylene Container with Screw Cap
Straight Polypropylene Container with Screw Cap
Fisherbrand
Straight Polypropylene Container with Screw Cap
Includes leakproof, polyethylene cap. Corning™ Gosselin™ Straight Polypropylene Container with Screw Cap is designed with a wide mouth to provide ample room for sample collection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityClosure Color
FKS15488834180 mLSterileRed
FIS07-202-026180 mLSterileRed
FKS15438844180 mLNon-sterileWhite
FKS1547880440 mLNon-sterileWhite
Catalog number FKS15488834
Price (SGD)
396.00
PK
Add to cart
Capacity (Metric):
180 mL
Sterility:
Sterile
Closure Color:
Red
Price (SGD)
396.00
PK
Add to cart
Designed for sample collection, storage and transportation to laboratories.
  • Polyethylene cap
  • Leakproof screw cap
  • Sterile or nonsterile versions available
Specifications
Capacity (Metric)180 mL
Capacity (English)6 oz.
ColorClear
DescriptionWide Mouth, Disposable
For Use With (Application)Sample collection
MaterialPolypropylene
No. per Pack264/Pk.
SterilitySterile
Closure ColorRed
Closure IncludedYes
GraduatedYes
ShapeCylindrical
Height (Metric)102 mm
Product TypeSample Container
Height (English)4 in.
Closure MaterialPolypropylene
Diameter (Metric)52 mm
Closure TypeScrew Cap
Unit SizePK