accuQuatro U Pack Cartridge
accuQuatro U Pack Cartridge
Actual product may vary
Fisherbrand
accuQuatro U Pack Cartridge
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
Have Questions?
Catalog NumberType
FKS15479709Cartridge, RO Feed
Catalog number FKS15479709
Price (SGD)
3,106.00
EA
Add to cart
Type:
Cartridge, RO Feed
Price (SGD)
3,106.00
EA
Add to cart
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
For Use With (Equipment)Accu100
TypeCartridge, RO Feed
Unit SizeEA