Straight Containers with Screw Cap
Straight Containers with Screw Cap
Actual product may vary
Fisherbrand
Straight Containers with Screw Cap
Collect, store, and transport samples. Gosselin™ Straight Containers with Screw Cap are CE marked for clinical use.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1546880440 mL
Catalog number FKS15468804
Price (SGD)
477.00
PK
Add to cart
Capacity (Metric):
40 mL
Price (SGD)
477.00
PK
Add to cart
  • Manufactured from natural polypropylene
  • Includes leakproof red LDPE screw cap
  • CE marking : CE, IVD
  • Food contact : Raw material is compliant with regulation EC Nº1935/2004
  • Sterility Assurance Level based on ISO 11137 : Nil
  • Packaged assembled
Specifications
Product TypeSpecimen Container
Closure IncludedYes
MaterialPolypropylene
ShapeStraight
SterilityNon-sterile
Capacity (Metric)40 mL
Closure ColorRed
Closure MaterialLDPE
Closure TypeScrew Cap
ColorClear
Diameter (Metric)30 mm
Height (Metric)70 mm
IncludesScrew cap
No. per Pack1000/Pk.
Unit SizePK