Isotemp™ Digital Dry Baths/Block Heaters
Isotemp™ Digital Dry Baths/Block Heaters
Actual product may vary
Isotemp™ Digital Dry Baths/Block Heaters
Interchangeable modular blocks, configurable for a range of applications, increase lab versatility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of PlacesPlug Type
FKS153979282CN, EU, UK
FKS153879281CN, EU, UK
Catalog number FKS15397928
Price (SGD)
1,897.00
EA
Add to cart
No. of Places:
2
Plug Type:
CN, EU, UK
Price (SGD)
1,897.00
EA
Add to cart
Fisherbrand™ Isotemp Digital Dry Baths/Block Heaters offer a range of configurations with interchangeable modular blocks for a variety of application needs.
Designed for basic to advanced applications with a maximum temperature of 130°C, a timer and available in 1, 2 and 4 block sizes.
  • Digital controls and display of time and temperature
  • Advanced internal temperature sensing probe for outstanding temperature accuracy and control
  • Precise temperature control with PID circuit
  • Temperature calibration enables to offset the temperature to desired value
  • Timer allows the user to accurately monitor the heating time
  • Wide range of interchangeable aluminum alloy heat blocks provide versatility and allow for easy cleaning and disinfecting
  • Powder-coated steel body construction ensures durability
  • Built-in over-temperature protection device warrants sample and user safety
  • Temperature deviation adjustment
Applications:
Sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation
Specifications
Certifications/ComplianceCE, cULus, RoHS
Controller TypePID
MaterialPowder-coated Steel
No. of Places2
Plug TypeCN, EU, UK
Relative Humidity<80%
Temperature Range (Metric)Ambient +5° to 130°C
TypeDry Bath Block
Fuse250V 5A
Heat Up Time≤ 20 min. 30° to 130°C
IncludesDry bath, CN, EU and UK plugs, block lifter
Temperature Accuracy≤ ± 0.5°C
Temperature Uniformity≤ ± 1°C
Timer0 to 99:59 min or continuous
Warranty2 Year, Replacement
Dimensions (L x W x H)12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm)
Length (English)12.5 in.
Length (Metric)318 mm
Width (English)7.9 in.
Height (English)3.9 in.
Height (Metric)100 mm
Weight (English)7.28 lb.
Weight (Metric)3.3 kg
Electrical Requirements200/240 V, 50/60 Hz (CN, EU, UK)
Voltage200/240 V