Black Polypropylene Screw Caps for Glass Specimen Vials
Actual product may vary
Fisherbrand
Black Polypropylene Screw Caps for Glass Specimen Vials
Screw securely into place to seal squat or tall form glass screw neck specimen vials. Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials are foil lined and available in sizes to fit a variety of tall or squat form vials.
Have Questions?
Catalog Number
Closure Size
FKS15384789
10 mm
Catalog number FKS15384789
Price (SGD)
57.00
PK
Add to cart
Closure Size:
10 mm
Price (SGD)
57.00
PK
Add to cart
Specifically designed for use with Fisherbrand™ Glass Screw Neck Specimen Vials
Polypropylene construction with foil lining
Fit both tall and squat form vials
Available in a variety of sizes to match vial specifications
Screw neck specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications