Sterilin™ Standard 90mm Petri Dishes
Sterilin™ Standard 90mm Petri Dishes
Actual product may vary
Fisherbrand
Sterilin™ Standard 90mm Petri Dishes
Culture microorganisms on solid media using these shallow dishes that maximize incubator space and feature a mirror finish that ensures high optical quality.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)No. per CaseSterilityVent
FKS1537036615.7 mm500Non-sterileNo
FKS1138927315.9 mm500Non-sterile1
FKS1130928316.1 mm500Non-sterile3
FKS1175925216.1 mm500Sterile3
FKS1173925215.7 mm500SterileNo
Catalog number FKS15370366
Price (SGD)
532.00
PK
Add to cart
Height (Metric):
15.7 mm
No. per Case:
500
Sterility:
Non-sterile
Vent:
No
Price (SGD)
532.00
PK
Add to cart
Culture microorganisms on solid media using Thermo Scientific™ Sterilin™ Standard 90mm Petri Dishes. These shallow dishes maximize incubator space and feature a mirror finish that ensures high optical quality.
NOT AVAILABLE IN AMERICA
Features:
  • Manufactured from non-cytotoxic virgin polystyrene
  • Ideal for use in automatic plate pourers
Available Vent Formats:
  • Triple vented—aids gaseous exchange; ideally suited for short-term work
  • Single vented—limits gaseous exchange, minimizes evaporation and dehydration; ideally suited for long-term work
  • Nonvented—most suitable for anaerobic and long-term work
Specifications
Certifications/ComplianceProduced and tested in accordance with the EN ISO 24998:2008 Petri dish standard which includes stringent dimensional controls.
DescriptionPetri dish, 90mm
MaterialPolystyrene
ShapeRound
SterilityNon-sterile
TypePetri Dish
No. per Case500
No. per Pack20
VentNo
Height (Metric)15.7 mm
Diameter (Metric)90 mm
Unit SizePK