AREX Hot Plate Stirrer with VTF, 370°C, Aluminum
AREX Hot Plate Stirrer with VTF, 370°C, Aluminum
Actual product may vary
Fisherbrand
AREX Hot Plate Stirrer with VTF, 370°C, Aluminum
Stirring Range: 1500 rpm; Size: 6.1 in. dia.
Have Questions?
Catalog NumberTop Plate Material
FKS15369244Aluminum
Catalog number FKS15369244
Price (SGD)
2,823.00
EA
Add to cart
Top Plate Material:
Aluminum
Price (SGD)
2,823.00
EA
Add to cart
Premium Quality Performance
  • Outstanding temp. uniformity with CerAlTop™ aluminum alloy heating plate
  • Excellent resistance to chemicals scratches and surface abrasions
  • Maximum temperature 370 °C
  • Accepts up to 20 liter flasks
  • Electronic speed regulation up to 1500 rpm
  • Excellent speed control, even at low speeds
  • SpeedServo™ ensures constant speed even when the viscosity changes
  • High power PCM-type driving magnet
Reliable Results
  • Connection for VTF Vertex digital thermoregulator for direct temperature control
  • VTF incorporates timer for unsupervised operation and increased productivity
Convenient Format
  • Available stand-alone or packaged with VTF Vertex digital thermoregulator
  • Control panel is IP 42 protected from liquid spills with run-off groove
  • Clear communication: front panel shows °C and rpm
  • LED interface enriched for alarms
Ergonomic Design
  • Inclined control panel for easy access and visibility
  • AluBlocks™ maximize temperature control & save handling time
  • Run different tubes by combining AluBlocks™
Specifications
Top Plate MaterialAluminum
Voltage230 V
IncludesWith VTF
Body MaterialEpoxy Painted Aluminum
Electrical Requirements230 V, 50 to 60 Hz, 630 w
ModelAREX
Sole FeaturesExcellent resistance to chemicals scratches and surface abrasions
Size6.1 in. dia.
Height (Metric)115 mm
Temperature (Metric)Ambient to 370°C
Frequency50/60 Hz
Width (Metric)165 mm
Wattage630 w
Weight (Metric)2.6 kg
TypeStirring Hotplate, Stirrer
Max. Container Size20 L
Stirring Range0 to 1500 rpm
Unit SizeEA