BoPET XRF Window Film
BoPET XRF Window Film
Actual product may vary
Fisherbrand
BoPET XRF Window Film
Thin window film for XRF liquid sample cups.
Have Questions?
Catalog NumberThicknessType
FKS153605620.25 milWindow Film Roll
Catalog number FKS15360562
Price (SGD)
520.00
EA
Add to cart
Thickness:
0.25 mil
Type:
Window Film Roll
Price (SGD)
520.00
EA
Add to cart
  • Heat resistant up to 100°C (212°F).
  • Unusually strong for its thickness.
  • Compatible with X-Cell™ sample cups.
  • Available in 0.12 mil (3 μm) and 0.25 mil (6 μm) thickness.
  • Cole-Parmer™ XRF window film serves as the sample window over our X-Cell sample cups, allowing for uniform transmission of x-rays while effectively containing the liquid sample.
  • BoPET film is heat resistant up to 100°C (212°F).
  • Unusually strong for its thickness, it is resistant to organic chemicals but vulnerable to strong acids and bases.
  • It may contain trace levels (ppm) of Ca, P, Fe, Cu, Zn, or Sb.
Specifications
For Use With (Equipment)XRF Spectrometers
TypeWindow Film Roll
DescriptionWindow Film Roll
Dimensions (L x W)300 ft. x 2.87 in.
For Use With (Application)Covers sample window
Length (English)300 ft.
Length (Metric)91.4 m
MaterialMylar
Width (English)2.87 in.
Quantity1 Roll
Thickness0.25 mil
Width (Metric)7.28 cm
Unit SizeEA