Samco™ Extra Long Transfer Pipettes
Samco™ Extra Long Transfer Pipettes
Actual product may vary
Samco™ Extra Long Transfer Pipettes
Samco™ Extra Long Transfer Pipettes
Samco™ Extra Long Transfer Pipettes
Fisherbrand
Samco™ Extra Long Transfer Pipettes
Perform safe and easy sampling from large containers using these extra-long transfer pipettes, perfect for research applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityPackaging TypeQuantity
FKS153578535.6 mLNon-sterileBulk400/Pk 8Pk/Cs 4000/Cs
FIS13-711-375.6 mLNon-sterileBulk400/Pk 8Pk/Cs 4000/Cs
262-1S5.6 mLSterileIndividually Wrapped400/Pk 8Pk/Cs 3200/Cs
FIS13-711-385.6 mLSterileIndividually Wrapped400/Pk 8Pk/Cs 3200/Cs
FIS13-711-4010.7 mLNon-sterileBulk100/Pk 10Pk/Cs 1000/Cs
FKS1165183923 mLNon-sterileWhite Box100/Pk.
FKS1134787323 mLSterileWhite Box20/Bag, 100/Pk.
Catalog number FKS15357853
Price (SGD)
79.00
PK
Add to cart
Volume (Metric):
5.6 mL
Sterility:
Non-sterile
Packaging Type:
Bulk
Quantity:
400/Pk 8Pk/Cs 4000/Cs
Price (SGD)
79.00
PK
Add to cart
Perfect for research applications, these Extra-Long Transfer Pipettes are designed with a built-in pipette bulb that eliminates the need for plugging, packaging, and autoclaving and prevents cross-contamination. They allow safe and easy sampling from large containers.
  • Excellent for research applications
  • General-purpose pipettes are ideal for routine sampling of liquids
  • Well suited for use with large containers
Specifications
Bulb Draw (Metric)2.3 mL
Diameter (Metric) Outer5.0 mm
MaterialLDPE
SterilityNon-sterile
Drops/mL25
GraduatedNo
Length (English)9 in.
Quantity400/Pk 8Pk/Cs 4000/Cs
Packaging TypeBulk
Volume (Metric)5.6 mL
Length (Metric)22.5 cm
TypeExtra Long Transfer Pipet