3-Part 50mL Luer Lock Syringes
3-Part 50mL Luer Lock Syringes
Actual product may vary
Fisherbrand
3-Part 50mL Luer Lock Syringes
Locks tip in place with integral Luer Lock. Terumo™ 3-Part 50mL Luer Lock Syringe includes a clear barrel and bold, black graduations for accurate measurements.
Have Questions?
Catalog NumberdimNeedleGauge
FKS15349067
Catalog number FKS15349067
Price (SGD)
73.00
PK
Add to cart
Price (SGD)
73.00
PK
Add to cart
  • Luer Lock
  • Concentric Tip
  • Graduations: 1ml
  • Sterile
  • Individually wrapped
  • Single-use
  • Latex-free
Specifications
ColorClear
DescriptionBlack graduations
Latex FreeYes
Product TypeSingle-use
DisposableSingle-use
Graduations1 mL
Quantity25/Pk.
Volume (Metric)50 mL
Connection TypeMale Luer Lock