Check Order Status
Labs Spectra/Por™ 7 50,000 D MWCO Standard RC Pre-Treated Dialysis Kits
Fisherbrand
Labs Spectra/Por™ 7 50,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Have Questions?
Catalog Number
Diameter (Metric)
Length (Metric)
FKS15340712
22 mm
3.7 mL/cm vol./length
Catalog number
FKS15340712
Price (SGD)
869.00
EA
Add to cart
Diameter (Metric):
22 mm
Length (Metric):
3.7 mL/cm vol./length
Price (SGD)
869.00
EA
Add to cart
Hydrophilic, symmetric porositiy
Equilibrated in water
Packaged wet in preservative solution of 0.05% sodium azide
For small experiments or qualifying a membrane for a particular application
Specifications
MWCO
50 kDa
Diameter (Metric)
22 mm
Length (Metric)
3.7 mL/cm vol./length
Quantity
1 m Tubing/Kit