Delrin™ Half-Size Test Tube Racks, 36 x 16 mm
Delrin™ Half-Size Test Tube Racks, 36 x 16 mm
Actual product may vary
Fisherbrand
Delrin™ Half-Size Test Tube Racks, 36 x 16 mm
Molded in a single, continuous piece. Delrin™ Half-Size Test Tube Racks, 36 x 16 mm are highly resistant to heat and chemical attack.
Have Questions?
Catalog NumberColor
FKS15340410White
Catalog number FKS15340410
Price (SGD)
41.00
EA
Add to cart
Color:
White
Price (SGD)
41.00
EA
Add to cart
  • Delrin™ fiberglass-reinforced polyoxymethylene (POM)
  • Offers the highest chemical and heat resistance (-40 to +140°C)
  • Autoclavable
  • Molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
Specifications
Product TypeTest Tube Rack
Array6 x 6
AutoclavableAutoclavable
ColorWhite
HoldsTubes (16 mm), Tubes (5 to 10 mL)
MaterialFiberglass-reinforced POM
Positions36
ShapeTeardrop-shaped Openings
Shelves2 Tiers
StackableYes, When Empty
SterilityNon-sterile
For Use With (Equipment)5 to 10 mL Test Tubes
No. per Case1
Quantity8 Pk.
Dimensions (L x W x H)127 x 127 x 70 mm
Length (Metric)127 mm
Length (English)5 in.
Width (Metric)127 mm
Width (English)5 in.
Height (Metric)70 mm
Height (English)2.8 in.
Diameter (Metric) Opening16 mm
Unit SizeEA