Check Order Status
Low-Form Weighing Bottles
Actual product may vary
Low-Form Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Low-Form Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
Have Questions?
Catalog Number
Capacity (Metric)
FKS15320895
45 mL
Catalog number
FKS15320895
Price (SGD)
707.00
PK
Add to cart
Capacity (Metric):
45 mL
Price (SGD)
707.00
PK
Add to cart
With interchangeable NS stopper or cap
With fused-in size (mm) and capacity (mL)
Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric)
45 mL
Form
Low
No. per Pack
1 Pack
Height (Metric)
30 mm
Product Type
Weighing Bottle
Diameter (Metric)
60 mm
Closure Type
Button Cover