Cryo Marker Pen Set
Cryo Marker Pen Set
Actual product may vary
Fisher Catalogue
Cryo Marker Pen Set
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS15289134Multi-color
4000221
also known as GT4000221
Multi-color
4000222
also known as GT4000222
Black
FIS11-679-117Black
Catalog number FKS15289134
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Multi-color
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
  • Multicolor set is available; includes 4 fine-tip pens in black, blue, green and red
  • Warranty varies by country, please contact us for details
Specifications
TypeCryo Marker Pen Set
Temperature Range-200°C, -328°F
ColorMulti-color
For Use With (Equipment)cryovials and boxes
HoldsVials
Unit SizePK