Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion
Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion
Actual product may vary
Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion
Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion
Fisherbrand
Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion
Easy-to-read partial immersion thermometers
Have Questions?
Catalog NumberTemperature Range (Metric)Temperature Range (English)SubdivisionLength (Metric) Overall
FKS15265053-35°C to +50°Cn.a.0.5°C300 mm
Catalog number FKS15265053
Price (SGD)
123.00
EA
Add to cart
Temperature Range (Metric):
-35°C to +50°C
Temperature Range (English):
n.a.
Subdivision:
0.5°C
Length (Metric) Overall:
300 mm
Price (SGD)
123.00
EA
Add to cart
  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Specifications
Certifications/ComplianceTraceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application)Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion TypePartial Immersion
Length (English) Overall12 in.
Length (Metric) Overall300 mm
MaterialGlass
Thermometer FillEnviro-Safe™ Liquid
Max. Temperature (Metric)50°C
Min. Temperature (Metric)-35°C
Temperature Range (English)n.a.
Temperature Range (Metric)-35°C to +50°C
Accuracy±1°C
Subdivision0.5°C
Immersion Length76 mm
Unit SizeEA