Check Order Status
Eutech Handheld Meters Calibration Solution, 84μ S / cm KCl
Element Search
Structure Search
Eutech Handheld Meters Calibration Solution, 84μ S / cm KCl
Have Questions?
Catalog number
FKS15264878
Price (SGD)
53.00
Price (SGD)
53.00
EA
Add to cart
Specifications
Recommended Storage
Store in cool location. Keep container tightly closed. Out of direct sun and away from heat source. Do not refrigerate.
Odor
Odorless
Density
1.0 g/cm³ at 25°C
Physical Form
Liquid
pH
∼7
View more
84μS/cm KCl calibration solution
Solutions packed in 480mL durable plastic bottles
Economical and convenient for laboratory use.