Eutech™ 5.0 mS/cm KCl Calibration Solution
Product Image
Eutech™ 5.0 mS/cm KCl Calibration Solution
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Have Questions?
Catalog NumberQuantity
FKS15263939480 mL
Catalog number FKS15263939
Price (SGD)
36.00
EA
Add to cart
Quantity:
480 mL
Price (SGD)
36.00
EA
Add to cart
Eutech™ 5.0 mS/cm KCl Calibration Solution is used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 480ml durable plastic bottles
  • Economical and convenient to use
Specifications
Solution TypeCalibration Solution
TypeConductivity Standard
Quantity480 mL
Unit SizeEA