Graduated Beakers with Handle, Embossed Scale
Graduated Beakers with Handle, Embossed Scale
Actual product may vary
Fisher Catalogue
Graduated Beakers with Handle, Embossed Scale
Manufactured from polypropylene. BRAND™ Graduated Beakers with Handle, Embossed Scale include an ergonomic handle for secure grip. Functional spout helps to minimize spills. Embossed scale will not diminish with autoclaving. Autoclavable at 121°C.
Have Questions?
Catalog NumberCapacity (Metric)
FKS152603545000 mL
Catalog number FKS15260354
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5000 mL
  • Manufactured from polypropylene.
  • Ergonomic handle provides secure grip.
  • Functional spout helps to minimize spills.
  • Embossed scale will not diminish with autoclaving.
  • Autoclavable at 121°C.
Specifications
Diameter (Metric)Top204 mm
Capacity (Metric)5000 mL
MaterialPolypropylene
Height (Metric)270 mm
Increments100 mL
Quantity6/Pk.
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceGMBH and CO KG
ScaleEmbossed Scale
Unit SizePK