DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Actual product may vary
DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Fisherbrand
DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Have Questions?
Catalog NumberSize
FKS15252119ST 24/29
Catalog number FKS15252119
Price (SGD)
61.00
PK
Add to cart
Size:
ST 24/29
Price (SGD)
61.00
PK
Add to cart
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
  • Made from polyethylene
  • Octagonal shaped head
Specifications
For Use With (Application)Storage of reagents, and chemicals
Quantity10/Pk.
SizeST 24/29
AutoclavableNot Autoclavable
MaterialPolyethylene
Unit SizePK