DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
Actual product may vary
DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
Fisherbrand
DURAN™ YOUTILITY™ Laboratory Bottle, GL 45
The smart ergonomic YOUTILITY™ laboratory bottle system helps to make laboratory work easier and safer.
Have Questions?
Catalog NumberCapacity (Metric)
FKS15223375250 mL
Catalog number FKS15223375
Price (SGD)
167.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
167.00
EA
Add to cart
The specially shaped gripping zones on both sides of the bottle permit easier and safer handling. The slimmer YOUTILITY™ bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators. The bottles can be conveniently stacked horizontally on their sides. A pre-defined labelling area is compatible with the dedicated self-adhesive YOUTILITY™ bottle labels. Sequentially sized bottles 125, 250, 500 and 1000 ml. The glass bottles are moulded from the tried and tested DURAN™ borosilicate 3.3, a pharmacopoeial Type 1 neutral glass. DURAN™ glass offers a very good chemical resistance and high temperature resistance.
  • Easier and safer handling with the specially shaped gripping zones on both sides of the bottle
  • Significantly faster opening or closing with the shortened bottle thread which remains fully compatible with GL 45 closures and other accessories
  • The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands
  • The pre-defined labelling areas are compatible with the dedicated DURAN™ self-adhesive YOUTILITY™ bottle labels for unambiguous bottle identification
  • The slimmer bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
  • The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal
  • Horizontal stacking is possible
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
Specifications
Capacity (Metric)250 mL
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sampling; Culture vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
MaterialBorosilicate Glass (3.3)
Quantity4/Pk.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Diameter (Metric)66 mm
Unit SizeEA