Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap
Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap
Actual product may vary
Fisherbrand
Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap
Manufactured from temperature and chemically resistant borosilicate glass. Pyrex™ Media-lab Bottles with Thermoplastic Polyester Cap includes chemically resistant PTFE sealing disc and pouring ring. White graduations and marking spot add easy volume assessment. Withstands hot air sterilization up to 180° C. Complies with ISO 4796.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS15221499100 mL45 mm
FKS1521149950 mL32 mm
Catalog number FKS15221499
Price (SGD)
728.00
Pack
Add to cart
Capacity (Metric):
100 mL
Closure Size:
45 mm
Price (SGD)
728.00
Pack
Add to cart
  • Manufactured from temperature and chemically resistant borosilicate glass
  • Bottles complete with thermoplastic polyester cap, PTFE sealing disc, and pouring ring
  • Withstands hot air sterilization up to 180° C
  • PTFE sealing disc resists chemical corrosion
  • White graduations and marking spot add easy volume assessment
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
Specifications
Capacity (Metric)100 mL
IncludesHigh temperature cap and pouring ring
MaterialBorosilicate Glass
Certifications/ComplianceISO 4796
Closure IncludedYes
Height (Metric)100 mm
Closure MaterialThermoplastic Polyester
Closure Size45 mm
Diameter (Metric)56 mm
Closure TypeCap
Unit SizePack