Check Order Status
Polypropylene Cryo Boxes
Actual product may vary
Fisherbrand
Polypropylene Cryo Boxes
Use these polypropylene boxes for low-temperature tube storage. Fisherbrand™ Polypropylene Cryo Boxes are ideal for sample storage and transportation.
Have Questions?
Change view
Catalog Number
Color
Type
Height (Metric)
FKS15220799
Black
Cryo Box, Without grid
52 mm
FKS10238380
Red
Cryo Box, With grid
52 mm
2 Options
Catalog number
FKS15220799
Price (SGD)
103.00
PK
Add to cart
Color:
Black
Type:
Cryo Box, Without grid
Height (Metric):
52 mm
Price (SGD)
103.00
PK
Add to cart
High quality polypropylene
Available in natural and other colors
Autoclavable
Temperature-resistant from -90 to +121°C
81 places, 9 x 9 grid, numerically coded
Robust hinge with safe snap-on lid
Stackable for safe transport
Drain bores at the bottom for dew liquids
Available with grid insert (5120-) or without grid insert (5220-)
Specifications
Closure Type
Snap-On Lid
Color
Black
Material
Polypropylene
Shape
Square
Type
Cryo Box, Without grid
For Use With (Application)
Safe Transport
Max. Temperature (Metric)
121°C
Min. Temperature (Metric)
-90°C
Quantity
5 Pack
Length (Metric)
133 mm
Width (Metric)
133 mm
Height (Metric)
52 mm