Glass Graduated Class AS Type 1 Pipets
Glass Graduated Class AS Type 1 Pipets
Actual product may vary
Fisherbrand
Glass Graduated Class AS Type 1 Pipets
Pipet liquids with precision. Fisherbrand™ graduated class AS type 1 pipets are made from soda lime glass. Type 1 pipets have the zero mark at the top.
Have Questions?
Catalog NumberVolume (Metric)
FKS1521793525 mL
Catalog number FKS15217935
Price (SGD)
37.00
PK
Add to cart
Volume (Metric):
25 mL
Price (SGD)
37.00
PK
Add to cart
Pipet liquids with precision. Fisherbrand™ graduated class AS type 1 pipets are made from soda lime glass. Type 1 pipets have the zero mark at the top.
  • Soda lime glass
  • Type 1
  • Blue graduations
  • Class AS
  • Batch Certificate of Compliance is downloadable 
Specifications
MaterialSoda Lime Glass
GraduatedYes
Quantity2 Pack
Volume (Metric)25 mL
TypePipet
Unit SizePK