Soda Lime Glass Pasteur Pipets
Soda Lime Glass Pasteur Pipets
Actual product may vary
Fisherbrand
Soda Lime Glass Pasteur Pipets
Constructed of Soda Lime Glass and complies with ISO 7712 standards. BRAND™ Soda Lime Glass Pasteur Pipets include a long, drawn-out capillary tip which is perfect for transferring small volumes of aqueous solutions. Suction end with constriction for a cotton plug allows filtering of small quantities.
Have Questions?
Catalog NumberLength (Metric)
FKS15212699225 mm
Catalog number FKS15212699
Price (SGD)
296.00
PK
Add to cart
Length (Metric):
225 mm
Price (SGD)
296.00
PK
Add to cart
  • Constructed of Soda Lime Glass.
  • Complies with ISO 7712 standards.
  • Long, drawn-out capillary tip is perfect for transferring small volumes of aqueous solutions.
  • Suction end with constriction for a cotton plug allows filtering of small quantities.
Specifications
Certifications/ComplianceISO 7712
Diameter (Metric) Outer7 mm
MaterialSoda Lime Glass
Quantity1000 Pack
Tip StyleCapillary
Volume (Metric)2 mL
Length (Metric)225 mm
TypePasteur Pipet
Unit SizePK