Sterile PES Syringe Filter
Sterile PES Syringe Filter
Actual product may vary
Sterile PES Syringe Filter
Designed for filtration and purification in a sterile environment. Fisherbrand™ Sterile PES Syringe Filter features a PES membrane in a polypropylene housing for purification of tissue culture solutions from a standard syringe. 33mm diameter membranes available with 0.2 or 0.45μm pore sizes.
Have Questions?
Catalog NumberPore Size
FKS152068690.2 μm
Catalog number FKS15206869
Price (SGD)
160.00
PK
Add to cart
Pore Size:
0.2 μm
Price (SGD)
160.00
PK
Add to cart
  • Ideal for purification of tissue culture solutions and buffers
  • High sample throughput with low protein binding
  • Hydrophilic 33mm polyethersulfone (PES) membrane
  • Sturdy, chemically resistant polypropylene housing
Specifications
ColorWhite
Material (Membrane)PES
Pore Size0.2 μm
SterilitySterile
For Use With (Application)Purification of tissue culture solutions, buffers
Housing MaterialPolypropylene
Quantity50/Pk.
TypeSyringe Filter
Diameter (Metric)33 mm
Unit SizePK