Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Actual product may vary
Fisherbrand
Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Epredia™ Shandon Straight Blunt/Sharp Dissecting Scissors are available in Premium and Standard grades.
Have Questions?
Catalog NumberLength (Metric)Grade
FKS1520229016.5 cmStandard
Catalog number FKS15202290
Price (SGD)
144.00
EA
Add to cart
Length (Metric):
16.5 cm
Grade:
Standard
Price (SGD)
144.00
EA
Add to cart
Consistent Quality
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship
Specifications
Product TypeDissecting Scissors
Blade StyleStraight
GradeStandard
Length (English)6.5 in.
Length (Metric)16.5 cm
Tip StyleBlunt/Sharp
Unit SizeEA