Conical Base Polypropylene Test Tubes
Conical Base Polypropylene Test Tubes
Actual product may vary
Fisherbrand
Conical Base Polypropylene Test Tubes
Compatible with standard rotors. Gosselin™ Conical Base Polypropylene Test Tubes come with green screw cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure IncludedSterility
FKS15193957YesNon-sterile
FKS11763274YesSterile
Catalog number FKS15193957
Price (SGD)
477.00
PK
Add to cart
Closure Included:
Yes
Sterility:
Non-sterile
Price (SGD)
477.00
PK
Add to cart
  • Sterile
  • Centrifuge tubes
  • With detachable sterility certificate
Specifications
Closure ColorBlue
ColorNatural
Capacity (Metric)50 mL
MaterialPolypropylene
No. per Pack600
PackagingBag
SterilityNon-sterile
Bottom ShapeConical
Closure IncludedYes
Diameter (Metric)29.2 mm
GraduatedYes
Product TypeCentrifuge Tube
Unit SizePK