Nalgene™ Pipet Cleaning Equipment Sets
Nalgene™ Pipet Cleaning Equipment Sets
Actual product may vary
Nalgene™ Pipet Cleaning Equipment Sets
Nalgene™ Pipet Cleaning Equipment Sets
Nalgene™ Pipet Cleaning Equipment Sets
Nalgene™ Pipet Cleaning Equipment Sets
Fisherbrand
Nalgene™ Pipet Cleaning Equipment Sets
Find everything you need to clean pipets in these pipet cleaning equipment sets, each conveniently packaged with basket, jar and washer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeFor Use With
FKS15192283FPipet, 32 in.
FIS15-350-111BCPipet, 24 in.
FKS10731421CPipet, 24 in.
FIS15-350-111CFPipet, 32 in.
Catalog number FKS15192283
Price (SGD)
5,089.00
PK
Add to cart
Size:
F
For Use With:
Pipet, 32 in.
Price (SGD)
5,089.00
PK
Add to cart
Acquire everything you need to clean pipets with the Thermo Scientific™ Nalgene™ Pipet Cleaning Equipment Sets. Each of these conveniently packaged sets features a basket, jar and washer. A larger size is available to accomodate up to 32 in. (81cm) long pipets.
Features:
  • All are made of rugged, high-density polyethylene
  • Components are also available separately except F size (sold only as a set)
Includes:
  • One pipet jar
  • One basket
  • One washer/rinser of size indicated
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
For Use WithPipet, 32 in.
Height (English)36 in.
Height (Metric)81 cm
SizeF
DescriptionPipet Cleaning Set F
MaterialHDPE
Includes
  • One pipet jar
  • One basket
  • One washer/rinser of size F
Unit SizePK