Check Order Status
Sterile PVDF Syringe Filter
Actual product may vary
Fisherbrand
Sterile PVDF Syringe Filter
Used for filtration of protein solutions.
Have Questions?
Catalog Number
Pore Size
FKS15191499
0.45 μm
Catalog number
FKS15191499
Price (SGD)
246.00
PK
Add to cart
Pore Size:
0.45 μm
Price (SGD)
246.00
PK
Add to cart
High sample throughput
Low protein binding
Hydrophilic in nature
Offers medium chemical resistance
Specifications
Material (Membrane)
PVDF
Pore Size
0.45 μm
Sterility
Sterile
For Use With (Application)
For a broad range of applications
Housing Material
Polypropylene
Quantity
50/Pk.
Type
Syringe Filter
Diameter (Metric)
33 mm